Keep my session open?
Ending In 
The session is expired
Twoja sesja wygasła. Zostałeś wylogowany dla własnego bezpieczeństwa.
Chcesz zalogować się jeszcze raz?

Mouse Anti-Human IgM Monoclonal Antibody [clone: P5E2]

Dostawca: Thermo Scientific
By ELISA and WB, MA5-14712 does not react with Human IgG or Human IgA. Product MA514729 is a smaller package size of MII0401 (formerly sold as a Seradyn product). Thermo Scientific Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Type: Secondary
Antigen: IgM
Clonality: Monoclonal
Clone: P5E2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human




Zaloguj się, aby zobaczyć swoje ceny i dostępność produktów.
  Pamiętaj mnie na tym urządzeniu
 

ZAMÓW

Rozwiń / Zwiń
Rozmiar Ilość Numer katalogowy VWR Ilość
100 ug 100 µG PIEAMA514729 Blocked
Blocked Ten produkt jest zablokowany. Prosimy, skontaktuj się z Działem Obsługi Klienta, aby uzyskać więcej informacji.
Wygeneruj kody kreskowe Format kodu kreskowego: 
Wygeneruj stronę katalogową PDF

SPECYFIKACJE

Antigen symbol IgM
Antigen name Immunoglobulin M
Reactivity Human
Conjugation Unconjugated
Clonality Monoclonal
Antibody type Secondary
Clone P5E2
Host Mouse
Isotype IgG1
Western Blot Yes
ELISA Yes
ImmunoPrecipitation Yes
Flow cytometry Yes
Antigen synonyms Immunoglobulin M, IgM
Stężenie 1 mg/ml
Immunogen Purified Human Serum IgM
Purification Ion-exchange chromatography
Form Liquid
Antibody fragment Whole Antibody
Storage Buffer 0.01M potassium phosphate, 0.135M NaCl, 1mg/ml BSA, 2.5mM KCl, 0.05% sodium azide
Storage temperature Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Shipping Temperature Ice
Wprowadź szczegóły

Select the article for which you are looking for Certificate

Podaj numer partii. To pole jest obowiązkowe. Wpisz "X" dla VWR Chemicals, jeśli nr partii nie jest znany.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

albo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Opis Numer katalogowy Availability Jednostka Twoja Cena Price Per Ilość