Keep my session open?
Ending In 
The session is expired
Twoja sesja wygasła. Zostałeś wylogowany dla własnego bezpieczeństwa.
Chcesz zalogować się jeszcze raz?

Anti-HBsAg Mouse Monoclonal Antibody [clone: P4A11]

Dostawca: Thermo Scientific
MA5-14731 targets Hepatitis B Surface Antigen in ELISA, IP, and RIA applications and shows reactivity with Virus samples. The MA5-14731 immunogen is hepatitis B Surface Antigen. Product MA514731 is a smaller package size of MIH9706 (formerly sold as a Seradyn product). Hepatitis B is an infectious illness caused by hepatitis B virus (HBV) which infects the liver of hominoidae, including humans, and causes an inflammation called hepatitis. Originally known as "serum hepatitis", the disease has caused epidemics in parts of Asia and Africa, and it is endemic in China. About a third of the world's population, more than 2 billion people, have been infected with the hepatitis B virus. This includes 350 million chronic carriers of the virus. Transmission of hepatitis B virus results from exposure to infectious blood or body fluids.

Type: Primary
Antigen: HBsAg
Clonality: Monoclonal
Clone: P4A11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Virus



Zaloguj się, aby zobaczyć swoje ceny i dostępność produktów.
  Pamiętaj mnie na tym urządzeniu
 

ZAMÓW

Rozwiń / Zwiń
Rozmiar Ilość Numer katalogowy VWR Ilość
100 ug 100 µG PIEAMA514731 Blocked
Blocked Ten produkt jest zablokowany. Prosimy, skontaktuj się z Działem Obsługi Klienta, aby uzyskać więcej informacji.
Wygeneruj kody kreskowe Format kodu kreskowego: 
Wygeneruj stronę katalogową PDF

SPECYFIKACJE

Antigen symbol HBsAg
Antigen name Hepatitis B Virus Surface Antigen (ay)
Reactivity Virus
Conjugation Unconjugated
Clonality Monoclonal
Antibody type Primary
Clone P4A11
Host Mouse
Isotype IgG2a
ELISA Yes
ImmunoPrecipitation Yes
Antigen synonyms Hepatitis B Surface Antigen, HepB surface antigen, HBsAg, HepB
Stężenie 1 mg/ml
Immunogen Hepatitis B Surface Antigen
Purification Protein A
Form Liquid
Storage Buffer 0.01M potassium phosphate, 2.5mM KCl, 0.135M NaCl, 1mg/ml BSA, 0.05% sodium azide
Storage temperature Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Shipping Temperature Ice
Wprowadź szczegóły

Select the article for which you are looking for Certificate

Podaj numer partii. To pole jest obowiązkowe. Wpisz "X" dla VWR Chemicals, jeśli nr partii nie jest znany.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

albo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Opis Numer katalogowy Availability Jednostka Twoja Cena Price Per Ilość