Keep my session open?
Ending In 
The session is expired
Twoja sesja wygasła. Zostałeś wylogowany dla własnego bezpieczeństwa.
Chcesz zalogować się jeszcze raz?

Anti-APOA1 Mouse Monoclonal Antibody [clone: 532]

Dostawca: Thermo Scientific
By sandwich ELISA, MA5-14732 can be used to detect Apo A-1 from serum samples. To increase sensitivity of sandwich ELISAs with MA5-14732, a biotinylated detection antibody followed by Streptavidin-HRP is recommended. By Western blot, MA5-14732 detects recombinant human Apo A-1, but not recombinant mouse Apo A-1. MA5-14732 is also recommended for detecting endogenous Apo A-1 by Western blot. By Western blot, MA5-14732 detects recombinant human Apo A-1, but not recombinant mouse Apo A-1. MA5-14732 is also recommended for detecting endogenous Apo A-1 by Western blot. Product MA514732 is a smaller package size of MIA1405 (formerly sold as a Seradyn product). This gene encodes apolipoprotein A-I, which is the major protein component of high density lipoprotein (HDL) in plasma. The protein promotes cholesterol efflux from tissues to the liver for excretion, and it is a cofactor for lecithin cholesterolacyltransferase (LCAT) which is responsible for the formation of most plasma cholesteryl esters. This gene is closely linked with two other apolipoprotein genes on chromosome 11. Defects in this gene are associated with HDL deficiencies, including Tangier disease, and with systemic non-neuropathic amyloidosis.

Type: Primary
Antigen: APOA1
Clonality: Monoclonal
Clone: 532
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human




 

ZAMÓW

Rozwiń / Zwiń
Rozmiar Ilość Numer katalogowy VWR Ilość
100 ug 100 µG PIEAMA514732 Blocked
Blocked Ten produkt jest zablokowany. Prosimy, skontaktuj się z Działem Obsługi Klienta, aby uzyskać więcej informacji.
Wygeneruj kody kreskowe Format kodu kreskowego: 
Wygeneruj stronę katalogową PDF

SPECYFIKACJE

Antigen symbol APOA1
Antigen name Apolipoprotein A1
Reactivity Human
Conjugation Unconjugated
Clonality Monoclonal
Antibody type Primary
Clone 532
Host Mouse
Isotype IgG1
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
ImmunoPrecipitation Yes
Antigen synonyms apoA-I, apo-AI
Gene ID 335
Stężenie 1 mg/ml
Immunogen Purified human plasma Apo A-1
Purification Protein A
Form Liquid
Storage buffer 0.01M potassium phosphate, 2.5mM KCl, 0.135M NaCl, 1mg/ml BSA, 0.05% sodium azide
Temperatura przechowywania Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Shipping temperature Ice
Wprowadź szczegóły

Select the article for which you are looking for Certificate

Podaj numer partii. To pole jest obowiązkowe. Wpisz "X" dla VWR Chemicals, jeśli nr partii nie jest znany.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

albo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Opis Numer katalogowy Availability Jednostka Twoja Cena Price Per Ilość