Keep my session open?
Ending In 
The session is expired
Twoja sesja wygasła. Zostałeś wylogowany dla własnego bezpieczeństwa.
Chcesz zalogować się jeszcze raz?

Anti-Ferritin Mouse Monoclonal Antibody [clone: 101]

Dostawca: Thermo Scientific
MA5-14733 detects Ferritin in ELISA, IP, IF, and RIA applications and shows reactivity to human samples. The MA5-14733 Immunogen is human liver ferritin. MA5-14733 detects ferritin which has a predicted molecular weight of approximately 19kDa. Product MA514733 is a smaller package size of MIF2501 (formerly sold as a Seradyn product). This gene encodes the light subunit of the ferritin protein. Ferritin is the major intracellular iron storage protein in prokaryotes and eukaryotes. It is composed of 24 subunits of the heavy and light ferritin chains. Variation in ferritin subunit composition may affect the rates of iron uptake and release in different tissues. A major function of ferritin is the storage of iron in a soluble and nontoxic state. Defects in this light chain ferritin gene are associated with several neurodegenerative diseases and hyperferritinemia-cataract syndrome. This gene has multiple pseudogenes.

Type: Primary
Antigen: Ferritin
Clonality: Monoclonal
Clone: 101
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human



Zaloguj się, aby zobaczyć swoje ceny i dostępność produktów.
  Pamiętaj mnie na tym urządzeniu
 

ZAMÓW

Rozwiń / Zwiń
Rozmiar Ilość Numer katalogowy VWR Ilość
100 ug 100 µG PIEAMA514733 Blocked
Blocked Ten produkt jest zablokowany. Prosimy, skontaktuj się z Działem Obsługi Klienta, aby uzyskać więcej informacji.
Wygeneruj kody kreskowe Format kodu kreskowego: 
Wygeneruj stronę katalogową PDF

SPECYFIKACJE

Antigen symbol Ferritin
Antigen name Ferritin
Reactivity Human
Conjugation Unconjugated
Clonality Monoclonal
Antibody type Primary
Clone 101
Host Mouse
Isotype IgG2a
ELISA Yes
ImmunoPrecipitation Yes
ImmunoFluorescence Yes
Antigen synonyms ferritin light chain, ferritin light polypeptide-like 2, ferritin L subunit, ferritin L-chain
Gene ID 2512
Stężenie 1 mg/ml
Immunogen Human liver ferritin
Purification Protein A
Form Liquid
Storage Buffer 0.01M potassium phosphate, 0.135M NaCl, 2.5mM KCl, 1mg/ml BSA, 0.05% sodium azide
Storage temperature Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Shipping Temperature Ice
Wprowadź szczegóły

Select the article for which you are looking for Certificate

Podaj numer partii. To pole jest obowiązkowe. Wpisz "X" dla VWR Chemicals, jeśli nr partii nie jest znany.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

albo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Opis Numer katalogowy Availability Jednostka Twoja Cena Price Per Ilość