Drukuj

Anti-Ferritin Mouse Monoclonal Antibody [clone: 513C10]

Dostawca: Thermo Fisher Scientific
PIEAMA514736EA 0 PLN
PIEAMA514736
Anti-Ferritin Mouse Monoclonal Antibody [clone: 513C10]
Przeciwciała
MA5-14736 detects Ferritin in ELISA, IP, IF, and RIA applications and shows reactivity to human samples. The MA5-14736 Immunogen is human liver ferritin. MA5-14736 detects ferritin which has a predicted molecular weight of approximately 19kDa. Product MA514736 is a smaller package size of MIF2504 (formerly sold as a Seradyn product). This gene encodes the light subunit of the ferritin protein. Ferritin is the major intracellular iron storage protein in prokaryotes and eukaryotes. It is composed of 24 subunits of the heavy and light ferritin chains. Variation in ferritin subunit composition may affect the rates of iron uptake and release in different tissues. A major function of ferritin is the storage of iron in a soluble and nontoxic state. Defects in this light chain ferritin gene are associated with several neurodegenerative diseases and hyperferritinemia-cataract syndrome. This gene has multiple pseudogenes.

Type: Primary
Antigen: Ferritin
Clonality: Monoclonal
Clone: 513C10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR