Keep my session open?
Ending In 
The session is expired
Twoja sesja wygasła. Zostałeś wylogowany dla własnego bezpieczeństwa.
Chcesz zalogować się jeszcze raz?

Anti-TSH Mouse Monoclonal Antibody [clone: Fc1003]

Dostawca: Thermo Scientific
MA5-14738 targets Thyroid Stimulating Hormone in ELISA, IP, and RIA applications and shows reactivity with Human samples. The MA5-14738 immunogen is human Thyroid Stimulating Hormone. MA5-14738 detects Thyroid Stimulating Hormone which has a predicted molecular weight of approximately 10 kDa. Product MA514738 is a smaller package size of MIT0406 (formerly sold as a Seradyn product). The four human glycoprotein hormones chorionic gonadotropin (CG), luteinizing hormone (LH), follicle stimulating hormone (FSH), and thyroid stimulating hormone (TSH) are dimers consisting of alpha and beta subunits that are associated noncovalently. The alpha subunits of these hormones are identical, however, their beta chains are unique and confer biological specificity. The protein encoded by this gene is the alpha subunit and belongs to the glycoprotein hormones alpha chain family.

Type: Primary
Antigen: TSH
Clonality: Monoclonal
Clone: Fc1003
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human




 

ZAMÓW

Rozwiń / Zwiń
Rozmiar Ilość Numer katalogowy VWR Ilość
100 ug 100 µG PIEAMA514738 Blocked
Blocked Ten produkt jest zablokowany. Prosimy, skontaktuj się z Działem Obsługi Klienta, aby uzyskać więcej informacji.
Wygeneruj kody kreskowe Format kodu kreskowego: 
Wygeneruj stronę katalogową PDF

SPECYFIKACJE

Antigen symbol TSH
Antigen name Thyroid Stimulating Hormone
Reactivity Human
Conjugation Unconjugated
Clonality Monoclonal
Antibody type Primary
Clone Fc1003
Host Mouse
Isotype IgG1
ELISA Yes
ImmunoPrecipitation Yes
Antigen synonyms chorionic gonadotrophin subunit alpha, follitropin alpha chain, lutropin alpha chain, follicle-stimulating hormone alpha chain, thyroid-stimulating hormone alpha chain, LSH-alpha, FSH-alpha, chorionic gonadotropin, glycoprotein hormones alpha chain, alpha polypeptide, follicle-stimulating hormone alpha subunit, anterior pituitary glycoprotein hormones common subunit alpha, thyrotropin alpha chain, choriogonadotropin alpha chain, TSH-alpha, luteinizing hormone alpha chain
Gene ID 1081
Stężenie 1 mg/ml
Immunogen Human Thyroid Stimulating Hormone
Purification Protein A
Form Liquid
Storage buffer 0.01M potassium phosphate, 1mg/ml BSA, 2.5mM KCl, 0.135M NaCl, 0.05% sodium azide
Temperatura przechowywania Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Shipping temperature Ice
Wprowadź szczegóły

Select the article for which you are looking for Certificate

Podaj numer partii. To pole jest obowiązkowe. Wpisz "X" dla VWR Chemicals, jeśli nr partii nie jest znany.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

albo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Opis Numer katalogowy Availability Jednostka Twoja Cena Price Per Ilość