Drukuj

Anti-APOB Mouse Monoclonal Antibody [clone: F2C13]

Dostawca: Thermo Fisher Scientific
PIEAMA514741EA 0 PLN
PIEAMA514741
Anti-APOB Mouse Monoclonal Antibody [clone: F2C13]
Przeciwciała
MA5-14741 targets Apolipoprotein B in ELISA, IP, and RIA applications and shows reactivity with Human samples. The MA5-14741 immunogen is purified human serum LDL. MA5-14741 detects Apolipoprotein B which has a predicted molecular weight of approximately 513 kDa. Product MA514741 is a smaller package size of MIA1610 (formerly sold as a Seradyn product). This gene product is the main apolipoprotein of chylomicrons and low density lipoproteins. It occurs in plasma as two main isoforms, apoB-48 and apoB-100: the former is synthesized exclusively in the gut and the latter in the liver. The intestinal and the hepatic forms of apoB are encoded by a single gene from a single, very long mRNA. The two isoforms share a common N-terminal sequence. The shorter apoB-48 protein is produced after RNA editing of the apoB-100 transcript at residue 2180 (CAA->UAA), resulting in the creation of a stop codon, and early translation termination. Mutations in this gene or its regulatory region cause hypobetalipoproteinemia, normotriglyceridemic hypobetalipoproteinemia, and hypercholesterolemia due to ligand-defective apoB, diseases affecting plasma cholesterol and apoB levels.

Type: Primary
Antigen: APOB
Clonality: Monoclonal
Clone: F2C13
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR