Keep my session open?
Ending In 
The session is expired
Twoja sesja wygasła. Zostałeś wylogowany dla własnego bezpieczeństwa.
Chcesz zalogować się jeszcze raz?

Anti-ALP Mouse Monoclonal Antibody [clone: P4A10E4]

Dostawca: Thermo Scientific
MA5-14742 targets Alkaline Phosphatase in ELISA, IHC, IP, and RIA applications and shows reactivity with Bovine samples. The MA5-14742 immunogen is purified Bovine Calf Intestine ALP. MA5-14742 detects Alkaline Phosphatase which has a predicted molecular weight of approximately 57 kDa. Product MA514742 is a smaller package size of MIA1703 (formerly sold as a Seradyn product). There are at least four distinct but related alkaline phosphatases: intestinal, placental, placental-like, and liver/bone/kidney (tissue non-specific). The intestinal alkaline phosphatase gene encodes a digestive brush-border enzyme. This enzyme is upregulated during small intestinal epithelial cell differentiation.

Type: Primary
Antigen: ALP
Clonality: Monoclonal
Clone: P4A10E4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine




 

ZAMÓW

Rozwiń / Zwiń
Rozmiar Ilość Numer katalogowy VWR Ilość
100 ug 100 µG PIEAMA514742 Blocked
Blocked Ten produkt jest zablokowany. Prosimy, skontaktuj się z Działem Obsługi Klienta, aby uzyskać więcej informacji.
Wygeneruj kody kreskowe Format kodu kreskowego: 
Wygeneruj stronę katalogową PDF

SPECYFIKACJE

Antigen symbol ALP
Antigen name Alkaline Phosphatase
Reactivity Bovine
Conjugation Unconjugated
Clonality Monoclonal
Antibody type Primary
Clone P4A10E4
Host Mouse
Isotype IgG1
ELISA Yes
ImmunoChemistry Yes
ImmunoPrecipitation Yes
Antigen synonyms placental alkaline phosphatase 1, placental type, alkaline phosphatase Regan isozyme, glycerophosphatase, PLAP-1, alkaline phosphomonoesterase, alkaline phosphatase, Regan isozyme
Gene ID 280993
Stężenie 1 mg/ml
Immunogen Purified Bovine Calf Intestine ALP
Purification Ion-exchange chromatography
Form Liquid
Storage buffer 0.01M potassium phosphate, 1mg/ml BSA, 0.135M NaCl, 2.5mM KCl, 0.05% sodium azide
Temperatura przechowywania Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Shipping temperature Ice
Wprowadź szczegóły

Select the article for which you are looking for Certificate

Podaj numer partii. To pole jest obowiązkowe. Wpisz "X" dla VWR Chemicals, jeśli nr partii nie jest znany.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

albo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Opis Numer katalogowy Availability Jednostka Twoja Cena Price Per Ilość