Keep my session open?
Ending In 
The session is expired
Twoja sesja wygasła. Zostałeś wylogowany dla własnego bezpieczeństwa.
Chcesz zalogować się jeszcze raz?

Anti-C Reactive Protein Mouse Monoclonal Antibody [clone: P4D7]

Dostawca: Thermo Scientific
MA5-14744 targets C Reactive Protein in ELISA, IP, and RIA applications and shows reactivity with Human samples. The MA5-14744 immunogen is purified human plasma CRP. MA5-14744 detects C Reactive Protein which has a predicted molecular weight of approximately 23 kDa. Product MA514744 is a smaller package size of MIC0501 (formerly sold as a Seradyn product). The protein encoded by this gene belongs to the pentaxin family. It is involved in several host defense related functions based on its ability to recognize foreign pathogens and damaged cells of the host and to initiate their elimination by interacting with humoral and cellular effector systems in the blood. Consequently, the level of this protein in plasma increases greatly during acute phase response to tissue injury, infection, or other inflammatory stimuli.

Type: Primary
Antigen: C Reactive Protein
Clonality: Monoclonal
Clone: P4D7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human




 

ZAMÓW

Rozwiń / Zwiń
Rozmiar Ilość Numer katalogowy VWR Ilość
100 ug 100 µG PIEAMA514744 Blocked
Blocked Ten produkt jest zablokowany. Prosimy, skontaktuj się z Działem Obsługi Klienta, aby uzyskać więcej informacji.
Wygeneruj kody kreskowe Format kodu kreskowego: 
Wygeneruj stronę katalogową PDF

SPECYFIKACJE

Antigen symbol C Reactive Protein
Antigen name C Reactive Protein
Reactivity Human
Conjugation Unconjugated
Clonality Monoclonal
Antibody type Primary
Clone P4D7
Host Mouse
Isotype IgG2a
ELISA Yes
ImmunoPrecipitation Yes
Gene ID 1401
Stężenie 1 mg/ml
Immunogen Purified human plasma CRP
Purification Ion-exchange chromatography
Form Liquid
Storage buffer 0.01M potassium phosphate, 1mg/ml BSA, 2.5mM KCl, 0.135M NaCl, 0.05% sodium azide
Temperatura przechowywania Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Shipping temperature Ice
Wprowadź szczegóły

Select the article for which you are looking for Certificate

Podaj numer partii. To pole jest obowiązkowe. Wpisz "X" dla VWR Chemicals, jeśli nr partii nie jest znany.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

albo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Opis Numer katalogowy Availability Jednostka Twoja Cena Price Per Ilość