Drukuj

Anti-Digoxin Mouse Monoclonal Antibody [clone: 25P1C9]

Dostawca: Thermo Fisher Scientific
PIEAMA514745EA 0 PLN
PIEAMA514745
Anti-Digoxin Mouse Monoclonal Antibody [clone: 25P1C9]
Przeciwciała
MA5-14745 targets Digoxin in Western Blot, ELISA, IP and RIA applications and shows reactivity with labeled oligonucleotides and chemical samples. Product MA514745 is a smaller package size of MID0301 (formerly sold as a Seradyn product). Digoxin, also known as digitalis, is a purified cardiac glycoside extracted from the foxglove plant, Digitalis lanata. Its corresponding aglycone is digoxigenin, and its acetyl derivative is acetyldigoxin. Digoxin is widely used in the treatment of various heart conditions, namely atrial fibrillation, atrial flutter and sometimes heart failure that cannot be controlled by other medication. Digoxin preparations are commonly marketed under the trade names Lanoxin, Digitek, and Lanoxicaps. Digoxin is also used as an affinity system for nonradioactive labeling and detection of proteins and nucleic acids.

Type: Primary
Antigen: Digoxin
Clonality: Monoclonal
Clone: 25P1C9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Chemical
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR