Keep my session open?
Ending In 
The session is expired
Twoja sesja wygasła. Zostałeś wylogowany dla własnego bezpieczeństwa.
Chcesz zalogować się jeszcze raz?

Anti-Digoxin Mouse Monoclonal Antibody [clone: 38G12]

Dostawca: Thermo Scientific
MA5-14746 targets Digoxin in Western Blot, ELISA, IP and RIA applications and shows reactivity with labeled oligonucleotides and chemical samples. Product MA514746 is a smaller package size of MID0302 (formerly sold as a Seradyn product). Digoxin, also known as digitalis, is a purified cardiac glycoside extracted from the foxglove plant, Digitalis lanata. Its corresponding aglycone is digoxigenin, and its acetyl derivative is acetyldigoxin. Digoxin is widely used in the treatment of various heart conditions, namely atrial fibrillation, atrial flutter and sometimes heart failure that cannot be controlled by other medication. Digoxin preparations are commonly marketed under the trade names Lanoxin, Digitek, and Lanoxicaps. Digoxin is also used as an affinity system for nonradioactive labeling and detection of proteins and nucleic acids.

Type: Primary
Antigen: Digoxin
Clonality: Monoclonal
Clone: 38G12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Chemical



Zaloguj się, aby zobaczyć swoje ceny i dostępność produktów.
  Pamiętaj mnie na tym urządzeniu
 

ZAMÓW

Rozwiń / Zwiń
Rozmiar Ilość Numer katalogowy VWR Ilość
100 ug 100 µG PIEAMA514746 Blocked
Blocked Ten produkt jest zablokowany. Prosimy, skontaktuj się z Działem Obsługi Klienta, aby uzyskać więcej informacji.
Wygeneruj kody kreskowe Format kodu kreskowego: 
Wygeneruj stronę katalogową PDF

SPECYFIKACJE

Antigen symbol Digoxin
Antigen name Digoxin
Reactivity Chemical
Conjugation Unconjugated
Clonality Monoclonal
Antibody type Primary
Clone 38G12
Host Mouse
Isotype IgG2a
Western Blot Yes
ELISA Yes
ImmunoPrecipitation Yes
Stężenie 1 mg/ml
Immunogen Digoxin-BSA
Purification Ion-exchange chromatography
Form Liquid
Storage Buffer 0.01M potassium phosphate, 0.135M NaCl, 1mg/ml BSA, 2.5mM KCl, 0.05% sodium azide
Storage temperature Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Shipping Temperature Ice
Wprowadź szczegóły

Select the article for which you are looking for Certificate

Podaj numer partii. To pole jest obowiązkowe. Wpisz "X" dla VWR Chemicals, jeśli nr partii nie jest znany.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

albo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Opis Numer katalogowy Availability Jednostka Twoja Cena Price Per Ilość