Drukuj

Anti-Estradiol Mouse Monoclonal Antibody [clone: AB4.2]

Dostawca: Thermo Fisher Scientific
PIEAMA514748EA 0 PLN
PIEAMA514748
Anti-Estradiol Mouse Monoclonal Antibody [clone: AB4.2]
Przeciwciała
MA5-14748 targets Estradiol in ELISA, IP, and RIA applications and shows reactivity with Chemical samples. The MA5-14748 immunogen is estradiol-6-CMO-KLH. Product MA514748 is a smaller package size of MIE0106 (formerly sold as a Seradyn product). Estradiol (17ß-estradiol) is a sex hormone. Estradiol is the predominant sex hormone present in females. It is also present in males, being produced as an active metabolic product of testosterone. It represents the major estrogen in humans. Estradiol has not only a critical impact on reproductive and sexual functioning, but also affects other organs including the bones.

Type: Primary
Antigen: Estradiol
Clonality: Monoclonal
Clone: AB4.2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Chemical
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR