Keep my session open?
Ending In 
The session is expired
Twoja sesja wygasła. Zostałeś wylogowany dla własnego bezpieczeństwa.
Chcesz zalogować się jeszcze raz?

Anti-Estradiol Mouse Monoclonal Antibody [clone: AB5.1]

Dostawca: Thermo Scientific
MA5-14749 targets Estradiol in ELISA, IP, and RIA applications and shows reactivity with Chemical samples. The MA5-14749 immunogen is estradiol-6-CMO-KLH. Product MA514749 is a smaller package size of MIE0108 (formerly sold as a Seradyn product). Estradiol (17ß-estradiol) is a sex hormone. Estradiol is the predominant sex hormone present in females. It is also present in males, being produced as an active metabolic product of testosterone. It represents the major estrogen in humans. Estradiol has not only a critical impact on reproductive and sexual functioning, but also affects other organs including the bones.

Type: Primary
Antigen: Estradiol
Clonality: Monoclonal
Clone: AB5.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Chemical




 

ZAMÓW

Rozwiń / Zwiń
Rozmiar Ilość Numer katalogowy VWR Ilość
100 ug 100 µG PIEAMA514749 Blocked
Blocked Ten produkt jest zablokowany. Prosimy, skontaktuj się z Działem Obsługi Klienta, aby uzyskać więcej informacji.
Wygeneruj kody kreskowe Format kodu kreskowego: 
Wygeneruj stronę katalogową PDF

SPECYFIKACJE

Antigen symbol Estradiol
Antigen name Estradiol
Reactivity Chemical
Conjugation Unconjugated
Clonality Monoclonal
Antibody type Primary
Clone AB5.1
Host Mouse
Isotype IgG1
ELISA Yes
ImmunoPrecipitation Yes
Antigen synonyms 17-Beta-Estradiol
Stężenie 1 mg/ml
Immunogen Estradiol-6-CMO-KLH
Purification Ion-exchange chromatography
Form Liquid
Storage buffer 0.01M potassium phosphate, 1mg/ml BSA, 2.5mM KCl, 0.135M NaCl, 0.05% sodium azide
Temperatura przechowywania Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Shipping temperature Ice
Wprowadź szczegóły

Select the article for which you are looking for Certificate

Podaj numer partii. To pole jest obowiązkowe. Wpisz "X" dla VWR Chemicals, jeśli nr partii nie jest znany.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

albo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Opis Numer katalogowy Availability Jednostka Twoja Cena Price Per Ilość