Keep my session open?
Ending In 
The session is expired
Twoja sesja wygasła. Zostałeś wylogowany dla własnego bezpieczeństwa.
Chcesz zalogować się jeszcze raz?

Anti-APOA5 Mouse Monoclonal Antibody [clone: 1G5G9 (c),2G1H11G9B3]

Dostawca: Thermo Scientific
MA5-14759 targets Apoa5 (ab) in WB applications and shows reactivity with Human samples. The MA5-14759 immnogen purified recombinant fragment of human APOA5 (Amino Acid: 20-363) expressed in E. Coli. The protein encoded by this gene is an apolipoprotein that plays an important role in regulating the plasma triglyceride levels, a major risk factor for coronary artery disease. It is a component of high density lipoprotein and is highly similar to a rat protein that is upregulated in response to liver injury. Mutations in this gene have been associated with hypertriglyceridemia and hyperlipoproteinemia type 5. This gene is located proximal to the apolipoprotein gene cluster on chromosome 11q23. Alternatively spliced transcript variants encoding the same protein have been identified.

Type: Primary
Antigen: APOA5
Clonality: Monoclonal
Clone: 1G5G9 (c),2G1H11G9B3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human




 

ZAMÓW

Rozwiń / Zwiń
Rozmiar Ilość Numer katalogowy VWR Ilość
100 ug 100 µG PIEAMA514759 Blocked
Blocked Ten produkt jest zablokowany. Prosimy, skontaktuj się z Działem Obsługi Klienta, aby uzyskać więcej informacji.
Wygeneruj kody kreskowe Format kodu kreskowego: 
Wygeneruj stronę katalogową PDF

SPECYFIKACJE

Antigen symbol APOA5
Antigen name Apolipoprotein A5
Reactivity Human
Conjugation Unconjugated
Clonality Monoclonal
Antibody type Primary
Clone 1G5G9 (c),2G1H11G9B3
Host Mouse
Isotype IgG1
Western blot Yes
Antigen synonyms APOAV, MGC126836, APOA5, APOA-V, MGC126838, RAP3
Gene ID 116519
Immunogen Purified recombinant fragment of human APOA5 (Amino Acid: 20-363)expressed in E. Coli.
Purification purified
Form Liquid
Storage buffer PBS, 0.03% sodium azide
Temperatura przechowywania Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Shipping temperature Ice
Wprowadź szczegóły

Select the article for which you are looking for Certificate

Podaj numer partii. To pole jest obowiązkowe. Wpisz "X" dla VWR Chemicals, jeśli nr partii nie jest znany.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

albo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Opis Numer katalogowy Availability Jednostka Twoja Cena Price Per Ilość