Keep my session open?
Ending In 
The session is expired
Twoja sesja wygasła. Zostałeś wylogowany dla własnego bezpieczeństwa.
Chcesz zalogować się jeszcze raz?

Anti-CD4 Mouse Monoclonal Antibody [clone: B486A1]

Dostawca: Thermo Scientific
MA5-14760 targets CD4 in FACS applications and shows reactivity with Human samples. The MA5-14760 immunogen is purified recombinant fragment of human CD4 expressed in E. Coli. This gene encodes a membrane glycoprotein of T lymphocytes that interacts with major histocompatibility complex class II antigenes and is also a receptor for the human immunodeficiency virus. This gene is expressed not only in T lymphocytes, but also in B cells, macrophages, and granulocytes. It is also expressed in specific regions of the brain. The protein functions to initiate or augment the early phase of T-cell activation, and may function as an important mediator of indirect neuronal damage in infectious and immune-mediated diseases of the central nervous system. Multiple alternatively spliced transcript variants encoding different isoforms have been identified in this gene.

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: B486A1
Conjugation: Unconjugated
Host: Mouse
Isotype: IgG1
Reactivity: Human

Zaloguj się, aby zobaczyć swoje ceny i dostępność produktów.
  Pamiętaj mnie na tym urządzeniu


Rozwiń / Zwiń
Rozmiar Ilość Numer katalogowy VWR Ilość
100 ug 100 µG PIEAMA514760 Blocked
Blocked Ten produkt jest zablokowany. Prosimy, skontaktuj się z Działem Obsługi Klienta, aby uzyskać więcej informacji.
Wygeneruj kody kreskowe Format kodu kreskowego: 
Wygeneruj stronę katalogową PDF


Antigen symbol CD4
Antigen name CD4 molecule
Reactivity Human
Conjugation Unconjugated
Clonality Monoclonal
Antibody type Primary
Clone B486A1
Host Mouse
Isotype IgG1
Flow cytometry Yes
Antigen synonyms CD4 antigen p55, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4, CD4 receptor
Gene ID 920
Immunogen Purified recombinant fragment of human CD4 expressed in E. Coli.
Purification purified
Form Liquid
Storage buffer PBS, 0.03% sodium azide
Temperatura przechowywania Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Shipping temperature Ice
Wprowadź szczegóły

Select the article for which you are looking for Certificate

Podaj numer partii. To pole jest obowiązkowe. Wpisz "X" dla VWR Chemicals, jeśli nr partii nie jest znany.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Opis Numer katalogowy Availability Jednostka Twoja Cena Price Per Ilość