Drukuj

Anti-KSHV K8a Mouse Monoclonal Antibody [clone: 8C12G10G1]

Dostawca: Thermo Fisher Scientific
PIEAMA514768EA 0 PLN
PIEAMA514768
Anti-KSHV K8a Mouse Monoclonal Antibody [clone: 8C12G10G1]
Przeciwciała
MA5-14768 targets KSHV K8a in IHC and WB applications and shows reactivity with Virus samples. The MA5-14768 immunogen is purified recombinant fragment of KSHV K8a expressed in E. Coli. Kaposi's sarcoma-associated herpesvirus (KSHV) belongs to the gamma-(2)-herpesvirus subfamily and has been closely linked to the Kaposi’s sarcoma, primary effusion lymphoma (PEL) and multicentric Castleman’s disease. The genome of KSHV is 165-170 kb and contains at least 88 open reading frames. KSHV K8 and K8.1 open reading frames are juxtaposed and span from nucleotide (nt) 74850 to 76695 of the virus genome. A K8 pre-mRNA overlaps the entire K8.1 coding region, and alternative splicing of KSHV K8 and K8.1 pre-mRNAs each produces three isoforms (α, β, and γ) of the mRNAs.

Type: Primary
Antigen: KSHV K8a
Clonality: Monoclonal
Clone: 8C12G10G1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Virus
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR