Keep my session open?
Ending In 
The session is expired
Twoja sesja wygasła. Zostałeś wylogowany dla własnego bezpieczeństwa.
Chcesz zalogować się jeszcze raz?

Anti-KSHV ORF45 Mouse Monoclonal Antibody [clone: 2D4A5]

Dostawca: Thermo Scientific
MA5-14769 targets KSHV ORF45 in IHC and WB applications and shows reactivity with Virus samples. The MA5-14769 immunogen is purified recombinant fragment of KSHV ORF45 expressed in E. Coli. Kaposi's sarcoma-associated herpesvirus (KSHV) belongs to the gamma-(2)-herpesvirus subfamily and has been closely linked to the Kaposi’s sarcoma, primary effusion lymphoma (PEL) and multicentric Castleman’s disease. The genome of KSHV is 165-170 kb and contains at least 88 open reading frames. ORF45 protein encoded by an immediate-early gene in the KSHV genome is characterized as a phosphorylated protein, and it is localized in the cytoplasm of infected cells. Studies have shown that ORF45 Protein interacted with cellular IRF-7 and blocked virus-mediated phosphorylation and nuclear translocation of IRF-7. In consequence, ORF45 efficiently inhibited virus-induced production of type I IFN. Zhu et al (2003) reported that the ORF45 Protein was associated with purified virions

Type: Primary
Antigen: KSHV ORF45
Clonality: Monoclonal
Clone: 2D4A5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Virus




 

ZAMÓW

Rozwiń / Zwiń
Rozmiar Ilość Numer katalogowy VWR Ilość
100 µL 100 µl PIEAMA514769 Blocked
Blocked Ten produkt jest zablokowany. Prosimy, skontaktuj się z Działem Obsługi Klienta, aby uzyskać więcej informacji.
Wygeneruj kody kreskowe Format kodu kreskowego: 
Wygeneruj stronę katalogową PDF

SPECYFIKACJE

Antigen symbol KSHV ORF45
Reactivity Virus
Conjugation Unconjugated
Clonality Monoclonal
Antibody type Primary
Clone 2D4A5
Host Mouse
Isotype IgG1
Western blot Yes
ImmunoChemistry Yes
Antigen synonyms Kaposi's sarcoma-associated herpesvirus
Format ascites
Immunogen Purified recombinant fragment of KSHV ORF45 expressed in E. Coli.
Form Liquid
Storage buffer ascites, 0.03% sodium azide
Temperatura przechowywania Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Shipping temperature Ice
Wprowadź szczegóły

Select the article for which you are looking for Certificate

Podaj numer partii. To pole jest obowiązkowe. Wpisz "X" dla VWR Chemicals, jeśli nr partii nie jest znany.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

albo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Opis Numer katalogowy Availability Jednostka Twoja Cena Price Per Ilość