Drukuj

Anti-KSHV ORF62 Mouse Monoclonal Antibody [clone: 5B7B6]

Dostawca: Thermo Fisher Scientific
PIEAMA514770EA 0 PLN
PIEAMA514770
Anti-KSHV ORF62 Mouse Monoclonal Antibody [clone: 5B7B6]
Przeciwciała
MA5-14770 targets KSHV ORF62 in IHC and WB applications and shows reactivity with Virus samples. The MA5-14770 immunogen is purified recombinant fragment of human KSHV ORF62 expressed in E. Coli. Kaposi's sarcoma-associated herpesvirus (KSHV) belongs to the gamma-(2)-herpesvirus subfamily and has been closely linked to the Kaposi’s sarcoma, primary effusion lymphoma (PEL) and multicentric Castleman’s disease. The genome of KSHV is 165-170 kb and contains at least 88 open reading frames. At least five major proteins are likely to be involved in the assembly of the HHV-8 capsid, including a protease (encoded by ORF17), the major capsid protein (encoded by ORF25), and three other smaller capsid proteins (encoded by ORF62, ORF26, and ORF65). Previous structural studies have shown that the HSV-1 triplex is a monomer of VP19c and a dimer of VP23 and that the HCMV triplex is similarly composed of a monomer and a dimer. By analogy, the KSHV triplexes are likely also composed of a monomer of the ORF62 protein and a dimer of the ORF26 protein, which are the respective homologs of VP19c and VP23.

Type: Primary
Antigen: KSHV ORF62
Clonality: Monoclonal
Clone: 5B7B6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Virus
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR