Keep my session open?
Ending In 
The session is expired
Twoja sesja wygasła. Zostałeś wylogowany dla własnego bezpieczeństwa.
Chcesz zalogować się jeszcze raz?

Anti-SARS CoV MP Mouse Monoclonal Antibody [clone: 2H2C4]

Dostawca: Thermo Scientific
MA5-14772 targets SARS-M in WB applications and shows reactivity with Virus samples. The MA5-14772 immunogen is purified recombinant fragment of SARS-m protein expressed in E. Coli. SARS (severe acute respiratory syndrome) is caused by a human coronavirus. Human coronaviruses are the major cause of upper respiratory tract illness, such as the common cold, in humans. Coronaviruses are positive-stranded RNA viruses, featuring the largest viral RNA genomes known to date (27-31 kb). The complete sequence of the SARS virus release the coronavirus contains 25 open reading frames. SARS-m is a membrane (M) protein which plays a the key player in virion assembly. One of its functions is to mediate the incorporation of the spikes into the viral envelope.

Type: Primary
Antigen: SARS CoV MP
Clonality: Monoclonal
Clone: 2H2C4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Virus




 

ZAMÓW

Rozwiń / Zwiń
Rozmiar Ilość Numer katalogowy VWR Ilość
100 µL 1 ml PIEAMA514772 Blocked
Blocked Ten produkt jest zablokowany. Prosimy, skontaktuj się z Działem Obsługi Klienta, aby uzyskać więcej informacji.
Wygeneruj kody kreskowe Format kodu kreskowego: 
Wygeneruj stronę katalogową PDF

SPECYFIKACJE

Antigen symbol SARS CoV MP
Antigen name SARS Coronavirus Matrix
Reactivity Virus
Conjugation Unconjugated
Clonality Monoclonal
Antibody type Primary
Clone 2H2C4
Host Mouse
Isotype IgG1
Western blot Yes
Antigen synonyms SARS Matrix Protein
Format tissue culture supernatant
Immunogen Purified recombinant fragment of SARS-m protein expressed in E. Coli.
Form Liquid
Storage buffer tissue culture supernatant, no preservative
Temperatura przechowywania Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Shipping temperature Ice
Wprowadź szczegóły

Select the article for which you are looking for Certificate

Podaj numer partii. To pole jest obowiązkowe. Wpisz "X" dla VWR Chemicals, jeśli nr partii nie jest znany.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

albo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Opis Numer katalogowy Availability Jednostka Twoja Cena Price Per Ilość