Keep my session open?
Ending In 
The session is expired
Twoja sesja wygasła. Zostałeś wylogowany dla własnego bezpieczeństwa.
Chcesz zalogować się jeszcze raz?

Anti-MME Mouse Monoclonal Antibody [clone: 2A1H5E1]

Dostawca: Thermo Scientific
MA5-15231 targets CD10 in IHC applications and shows reactivity with Human samples. The MA5-15231 immunogen is purified recombinant fragment of human CD-10 expressed in E. Coli. CD10 is a 100kDa glycoprotein, also designated Common Acute Lymphocytic Leukemia Antigen (CALLA). It is a cell surface enzyme with neutral metalloendopeptidase activity which inactivates a variety of biologically active peptides. CD10 is expressed on the cells of lymphoblastic, Burkitt’s, and follicular germinal center lymphomas, and on cells from patients with chronic myelocytic leukemia (CML). It is also expressed on the surface of normal early lymphoid progenitor cells, immature B cells within adult bone marrow and germinal center B cells within lymphoid tissue. CD10 is also present on breast myoepithelial cells, bile canaliculi, fibroblasts, with especially high expression on the brush border of kidney and gut epithelial cells.

Type: Primary
Antigen: MME
Clonality: Monoclonal
Clone: 2A1H5E1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human



Zaloguj się, aby zobaczyć swoje ceny i dostępność produktów.
  Pamiętaj mnie na tym urządzeniu
 

ZAMÓW

Rozwiń / Zwiń
Rozmiar Ilość Numer katalogowy VWR Ilość
100 ug 100 µG PIEAMA515231 Blocked
Blocked Ten produkt jest zablokowany. Prosimy, skontaktuj się z Działem Obsługi Klienta, aby uzyskać więcej informacji.
Wygeneruj kody kreskowe Format kodu kreskowego: 
Wygeneruj stronę katalogową PDF

SPECYFIKACJE

Antigen symbol MME
Antigen name Neprilysin
Reactivity Human
Conjugation Unconjugated
Clonality Monoclonal
Antibody type Primary
Clone 2A1H5E1
Host Mouse
Isotype IgG2b
ImmunoChemistry Yes
Antigen synonyms SFE, atriopeptidase, membrane metallo-endopeptidase, NEP, CD10, membrane metallo-endopeptidase neutral endopeptidase, common acute lymphocytic leukemia antigen, enkephalinase, CALLA
Gene ID 4311
Immunogen Purified recombinant fragment of human CD-10 expressed in E. Coli.
Purification purified
Form Liquid
Storage Buffer PBS, 0.03% sodium azide
Storage temperature Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Shipping Temperature Ice
Wprowadź szczegóły

Select the article for which you are looking for Certificate

Podaj numer partii. To pole jest obowiązkowe. Wpisz "X" dla VWR Chemicals, jeśli nr partii nie jest znany.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

albo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Opis Numer katalogowy Availability Jednostka Twoja Cena Price Per Ilość