Drukuj

Anti-CD79a Mouse Monoclonal Antibody [clone: 2F11]

Dostawca: Thermo Fisher Scientific
PIEAMA515234EA 0 PLN
PIEAMA515234
Anti-CD79a Mouse Monoclonal Antibody [clone: 2F11]
Przeciwciała
MA5-15234 targets Neurofilament (200kDa and 68kDa) in IF, IHC (P), IP, and WB applications and shows reactivity with Feline, Human, mouse, Rabbit, and Rat samples. The MA5-15234 immunogen is neurofilament purified from human brain. Neurofilaments are the intermediate filaments of neurons and their processes. They are protein triplets composed of three major subunits of MW 68kDa, 155kDa, and 200kDa. Neurofilaments are expressed in tumors of neural origin, or displaying neuronal differentiation, such as neuroblastoma, medulloblastoma, and retinoblastoma

Type: Primary
Antigen: CD79a
Clonality: Monoclonal
Clone: 2F11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human, Feline, Mouse, Rabbit, Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR