Drukuj

Anti-MLANA Mouse Monoclonal Antibody [clone: M2-7C10]

Dostawca: Thermo Fisher Scientific
PIEAMA515237EA 0 PLN
PIEAMA515237
Anti-MLANA Mouse Monoclonal Antibody [clone: M2-7C10]
Przeciwciała
MA5-15237 targets MART-1 in FACS, IHC (P), IP, and WB applications and shows reactivity with Human samples. The MA5-15237 immunogen is recombinant hMART-1 protein. MART-1 (Melanoma Antigen Recognized by T cells 1) or Melan-Ais a newly identified melanocyte differentiation antigen recognized by autologous cytotoxic T lymphocytes. Seven other melanoma associated antigens recognized by autologous cytotoxic T cells include MAGE-1, MAGE-3, tyrosinase, gp100, gp75, BAGE-1, and GAGE-1. Subcellular fractionation shows that MART-1 is present in melanosomes and endoplasmic reticulum.

Type: Primary
Antigen: MLANA
Clonality: Monoclonal
Clone: M2-7C10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b, kappa
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR