Drukuj

Anti-P53 Mouse Monoclonal Antibody [clone: PAb 240]

Dostawca: Thermo Fisher Scientific
PIEAMA515244EA 0 PLN
PIEAMA515244
Anti-P53 Mouse Monoclonal Antibody [clone: PAb 240]
Przeciwciała
MA5-15244 targets p53 in FACS, IHC (P), IP, and WB applications and shows reactivity with Human samples. The MA5-15244 immunogen is mouse p53 protein corresponding to aa14-289. This gene encodes a tumor suppressor protein containing transcriptional activation, DNA binding, and oligomerization domains. The encoded protein responds to diverse cellular stresses to regulate expression of target genes, thereby inducing cell cycle arrest, apoptosis, senescence, DNA repair, or changes in metabolism. Mutations in this gene are associated with a variety of human cancers, including hereditary cancers such as Li-Fraumeni syndrome. Alternative splicing of this gene and the use of alternate promoters result in multiple transcript variants and isoforms. Additional isoforms have also been shown to result from the use of alternate translation initiation codons

Type: Primary
Antigen: P53
Clonality: Monoclonal
Clone: PAb 240
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rhesus Monkey
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR