Drukuj

Anti-GFP Mouse Monoclonal Antibody [clone: GF28R]

Dostawca: Thermo Fisher Scientific
PIEAMA515256EA 0 PLN
PIEAMA515256
Anti-GFP Mouse Monoclonal Antibody [clone: GF28R]
Przeciwciała
MA5-15256 recognizes native and denatured forms of GFP (Green Fluorescent Protein) and its many popular variants: EGFP, YFP, EYFP, and CFP. MA5-15256 has successfully been used in immunoprecipitation, ELISA, Western blotting, and immunofluorescence applications. MA5-15256 may be stored at 4ºC short term. Add 0.05% sodium azide if desired. For long term storage, store at -20ºC, avoiding freeze/thaw cycles. Epitope tags provide a method to localize gene products in a variety of cell types, study the topology of proteins and protein complexes, identify associated proteins, and characterize newly identified, low abundance or poorly immunogenic proteins when protein specific antibodies are not available. Green Fluorescent Protein (GFP) has quickly become a powerful research tool for assessing gene expression and subcellular protein distribution in fixed or living cells. GFP is excited by and brightly fluoresces when exposed to UV or blue light. This feature makes it ideal as a marker for use in fluorescence microscopy, cytometry, tagging fusion proteins, and assaying transcriptional regulation from gene promoters in vivo. Numerous GFP variants with enhanced and shifted emission spectra (blue, green, and yellow) have been developed through amino acid substitutions at specific residues.

Type: Primary
Antigen: GFP
Clonality: Monoclonal
Clone: GF28R
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Tag
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR