Keep my session open?
Ending In 
The session is expired
Twoja sesja wygasła. Zostałeś wylogowany dla własnego bezpieczeństwa.
Chcesz zalogować się jeszcze raz?

Anti-Mouse TUG Mouse Monoclonal Antibody [clone: 4A11A6G11]

Dostawca: Thermo Scientific
MA5-15269 targets Mouse TUG in WB applications and shows reactivity with mouse samples. The MA5-15269 immunogen is purified recombinant fragment of TUG expressed in E. Coli. This gene is a candidate gene for alveolar soft part sarcoma . It has been found that this gene is fused with transcription factor TFE3 gene in ASPS and also in renal cell carcinomas. Several alternatively spliced transcript variants of this gene have been described, but their full length nature has not been determined.

Type: Primary
Antigen: Mouse TUG
Clonality: Monoclonal
Clone: 4A11A6G11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse




 

ZAMÓW

Rozwiń / Zwiń
Rozmiar Ilość Numer katalogowy VWR Ilość
100 ug 100 µG PIEAMA515269 Blocked
Blocked Ten produkt jest zablokowany. Prosimy, skontaktuj się z Działem Obsługi Klienta, aby uzyskać więcej informacji.
Wygeneruj kody kreskowe Format kodu kreskowego: 
Wygeneruj stronę katalogową PDF

SPECYFIKACJE

Antigen symbol Mouse TUG
Antigen name Mouse TUG
Reactivity Mouse
Conjugation Unconjugated
Clonality Monoclonal
Antibody type Primary
Clone 4A11A6G11
Host Mouse
Isotype IgG1
Western blot Yes
Antigen synonyms ASPCR1, ASPL, ASPS, RCC17, TUG, UBXD9, UBXN9
Gene ID 68938
Immunogen Purified recombinant fragment of TUG expressed in E. Coli.
Purification purified
Form Liquid
Storage buffer PBS, 0.03% sodium azide
Temperatura przechowywania Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Shipping temperature Ice
Wprowadź szczegóły

Select the article for which you are looking for Certificate

Podaj numer partii. To pole jest obowiązkowe. Wpisz "X" dla VWR Chemicals, jeśli nr partii nie jest znany.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

albo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Opis Numer katalogowy Availability Jednostka Twoja Cena Price Per Ilość
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping