Drukuj

Anti-EPHA1 Mouse Monoclonal Antibody [clone: 5D2]

Dostawca: Thermo Fisher Scientific
PIEAMA515271EA 0 PLN
PIEAMA515271
Anti-EPHA1 Mouse Monoclonal Antibody [clone: 5D2]
Przeciwciała
MA5-15271 targets EphA1 in IHC and WB applications and shows reactivity with Human samples. The MA5-15271 immunogen is purified recombinant fragment of EphA1 expressed in E. Coli. MA5-15271 detects EphA1 which has a predicted molecular weight of approximately 108kDa. This gene belongs to the ephrin receptor subfamily of the protein-tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediating developmental events, particularly in the nervous system. Receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. The ephrin receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. This gene is expressed in some human cancer cell lines and has been implicated in carcinogenesis.

Type: Primary
Antigen: EPHA1
Clonality: Monoclonal
Clone: 5D2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR