Keep my session open?
Ending In 
The session is expired
Twoja sesja wygasła. Zostałeś wylogowany dla własnego bezpieczeństwa.
Chcesz zalogować się jeszcze raz?

Anti-Calcyclin Mouse Monoclonal Antibody [clone: 7D11]

Dostawca: Thermo Scientific
MA5-15272 targets Calcyclin in IHC and WB applications and shows reactivity with Human samples. The MA5-15272 immunogen is purified recombinant fragment of calcyclin expressed in E. Coli. Calcyclin encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein may function in stimulation of Ca2+-dependent insulin release, stimulation of prolactin secretion, and exocytosis. Chromosomal rearrangements and altered expression of this gene have been implicated in melanoma.

Type: Primary
Antigen: Calcyclin
Clonality: Monoclonal
Clone: 7D11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human



Zaloguj się, aby zobaczyć swoje ceny i dostępność produktów.
  Pamiętaj mnie na tym urządzeniu
 

ZAMÓW

Rozwiń / Zwiń
Rozmiar Ilość Numer katalogowy VWR Ilość
100 ug 100 µG PIEAMA515272 Blocked
Blocked Ten produkt jest zablokowany. Prosimy, skontaktuj się z Działem Obsługi Klienta, aby uzyskać więcej informacji.
Wygeneruj kody kreskowe Format kodu kreskowego: 
Wygeneruj stronę katalogową PDF

SPECYFIKACJE

Antigen symbol Calcyclin
Antigen name Calcyclin
Reactivity Human
Conjugation Unconjugated
Clonality Monoclonal
Antibody type Primary
Clone 7D11
Host Mouse
Isotype IgG1
Western Blot Yes
ImmunoChemistry Yes
Antigen synonyms 5B10, CACY, S100A6, 2A9, Calcyclin, PRA, CABP
Gene ID 27101
Immunogen Purified recombinant fragment of calcyclin expressed in E. Coli.
Purification purified
Form Liquid
Storage Buffer PBS, 0.03% sodium azide
Storage temperature Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Shipping Temperature Ice
Wprowadź szczegóły

Select the article for which you are looking for Certificate

Podaj numer partii. To pole jest obowiązkowe. Wpisz "X" dla VWR Chemicals, jeśli nr partii nie jest znany.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

albo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Opis Numer katalogowy Availability Jednostka Twoja Cena Price Per Ilość