Keep my session open?
Ending In 
The session is expired
Twoja sesja wygasła. Zostałeś wylogowany dla własnego bezpieczeństwa.
Chcesz zalogować się jeszcze raz?

Anti-HPRT1 Mouse Monoclonal Antibody [clone: 1F8D11]

Dostawca: Thermo Scientific
MA5-15274 targets HPRT in WB applications and shows reactivity with Human samples. The MA5-15274 immunogen is purified recombinant fragment of HPRT expressed in E. Coli. The HPRT1 gene provides instructions for making an enzyme called hypoxanthine phosphoribosyltransferase 1. This enzyme allows cells to recycle purines, some of the building blocks of DNA and its chemical cousin RNA.The enzyme hypoxanthine-guanine phosphoribosyltrasferase (E.C.2.4.2.8., HPRT) plays a crucial role in uric acid synthesis and purine metabolism.This enzyme catalyzes the conversion of hypoxanthine and guanine to inosine monophosphate (IMP) and guanosine monophosphate (GMP), respectively, and uses phosphoribosylpyrophosphate (PRPP) as a cosubstrate and as a source of energy . This pathway is also known as the purine salvage pathway because it allows cells to reuse purine compounds to build DNA and RNA.

Type: Primary
Antigen: HPRT1
Clonality: Monoclonal
Clone: 1F8D11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human



Zaloguj się, aby zobaczyć swoje ceny i dostępność produktów.
  Pamiętaj mnie na tym urządzeniu
 

ZAMÓW

Rozwiń / Zwiń
Rozmiar Ilość Numer katalogowy VWR Ilość
100 µL 100 µl PIEAMA515274 Blocked
Blocked Ten produkt jest zablokowany. Prosimy, skontaktuj się z Działem Obsługi Klienta, aby uzyskać więcej informacji.
Wygeneruj kody kreskowe Format kodu kreskowego: 
Wygeneruj stronę katalogową PDF

SPECYFIKACJE

Antigen symbol HPRT1
Antigen name Hypoxanthine phosphoribosyltransferase 1
Reactivity Human
Conjugation Unconjugated
Clonality Monoclonal
Antibody type Primary
Clone 1F8D11
Host Mouse
Isotype IgG2b
Western Blot Yes
Antigen synonyms HGPRTase, HGPRT, HPRT, HPRT1, Hypoxanthine-guanine phosphoribosyltransferase
Gene ID 3251
Format ascites
Immunogen Purified recombinant fragment of HPRT expressed in E. Coli.
Form Liquid
Storage Buffer ascites, 0.03% sodium azide
Storage temperature Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Shipping Temperature Ice
Wprowadź szczegóły

Select the article for which you are looking for Certificate

Podaj numer partii. To pole jest obowiązkowe. Wpisz "X" dla VWR Chemicals, jeśli nr partii nie jest znany.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

albo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Opis Numer katalogowy Availability Jednostka Twoja Cena Price Per Ilość