Keep my session open?
Ending In 
The session is expired
Twoja sesja wygasła. Zostałeś wylogowany dla własnego bezpieczeństwa.
Chcesz zalogować się jeszcze raz?

Anti-NPT Mouse Monoclonal Antibody [clone: 4B4D1]

Dostawca: Thermo Scientific
MA5-15275 targets NPT in WB applications and shows reactivity with Human samples. The MA5-15275 immunogen is purified recombinant fragment of NPT expressed in E. Coli. Neomycin phosphotransferase II (nptII) gene is used in selection of transformed organisms. It was initially isolated from the transposon Tn5 that was present in the bacterium strain Escherichia coli K12. The gene codes for the aminoglycoside 3'-phosphotransferase (denoted aph(3')-II or NPTII) enzyme. NPTII is probably the most widely used selectable marker for plant transformation. It is also used in gene expression and regulation studies in different organisms in part because N-terminal fusions can be constructed that retain enzymatic activity. In animal cells, G418 and neomycin are used as selectable agents.

Type: Primary
Antigen: NPT
Clonality: Monoclonal
Clone: 4B4D1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Bacteria




 

ZAMÓW

Rozwiń / Zwiń
Rozmiar Ilość Numer katalogowy VWR Ilość
100 µL 100 µl PIEAMA515275 Blocked
Blocked Ten produkt jest zablokowany. Prosimy, skontaktuj się z Działem Obsługi Klienta, aby uzyskać więcej informacji.
Wygeneruj kody kreskowe Format kodu kreskowego: 
Wygeneruj stronę katalogową PDF

SPECYFIKACJE

Antigen symbol NPT
Antigen name Sodium/phosphate cotransporter
Reactivity Bacteria
Conjugation Unconjugated
Clonality Monoclonal
Antibody type Primary
Clone 4B4D1
Host Mouse
Isotype IgG2a
Western blot Yes
Format ascites
Immunogen Purified recombinant fragment of NPT expressed in E. Coli.
Form Liquid
Storage buffer ascites, 0.03% sodium azide
Temperatura przechowywania Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Shipping temperature Ice
Wprowadź szczegóły

Select the article for which you are looking for Certificate

Podaj numer partii. To pole jest obowiązkowe. Wpisz "X" dla VWR Chemicals, jeśli nr partii nie jest znany.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

albo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Opis Numer katalogowy Availability Jednostka Twoja Cena Price Per Ilość