Drukuj

Anti-LAL Mouse Monoclonal Antibody [clone: 9G7F12,7G6D7]

Dostawca: Thermo Fisher Scientific
PIEAMA515278EA 0 PLN
PIEAMA515278
Anti-LAL Mouse Monoclonal Antibody [clone: 9G7F12,7G6D7]
Przeciwciała
MA5-15278 targets LAL in WB applications and shows reactivity with Human samples. The MA5-15278 immunogen is purified recombinant fragment of LAL expressed in E. Coli. This gene encodes lipase A, the lysosomal acid lipase . This enzyme functions in the lysosome to catalyze the hydrolysis of cholesteryl esters and triglycerides. Mutations in this gene can result in Wolman disease and cholesteryl ester storage disease. Alternatively spliced transcript variants encoding the same protein have been found for this gene.

Type: Primary
Antigen: LAL
Clonality: Monoclonal
Clone: 9G7F12,7G6D7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR