Keep my session open?
Ending In 
The session is expired
Twoja sesja wygasła. Zostałeś wylogowany dla własnego bezpieczeństwa.
Chcesz zalogować się jeszcze raz?

Anti-PTK2 Mouse Monoclonal Antibody [clone: 4A9D6]

Dostawca: Thermo Scientific
MA5-15282 targets FAK in WB applications and shows reactivity with Human samples. The MA5-15282 immunogen is purified recombinant fragment of FAK expressed in E. Coli. This gene encodes a cytoplasmic protein tyrosine kinase which is found concentrated in the focal adhesions that form between cells growing in the presence of extracellular matrix constituents. The encoded protein is a member of the FAK subfamily of protein tyrosine kinases but lacks significant sequence similarity to kinases from other subfamilies. Activation of this gene may be an important early step in cell growth and intracellular signal transduction pathways triggered in response to certain neural peptides or to cell interactions with the extracellular matrix. Several transcript variants encoding different isoforms have been found for this gene, but the full-length natures of only three of them have been determined.

Type: Primary
Antigen: PTK2
Clonality: Monoclonal
Clone: 4A9D6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human




 

ZAMÓW

Rozwiń / Zwiń
Rozmiar Ilość Numer katalogowy VWR Ilość
100 µL 100 µl PIEAMA515282 Blocked
Blocked Ten produkt jest zablokowany. Prosimy, skontaktuj się z Działem Obsługi Klienta, aby uzyskać więcej informacji.
Wygeneruj kody kreskowe Format kodu kreskowego: 
Wygeneruj stronę katalogową PDF

SPECYFIKACJE

Antigen symbol PTK2
Antigen name Protein Tyrosine Kinase 2
Reactivity Human
Conjugation Unconjugated
Clonality Monoclonal
Antibody type Primary
Clone 4A9D6
Host Mouse
Isotype IgG
Western blot Yes
Antigen synonyms PTK2, pp125FAK, FAK, FAK1, FADK
Gene ID 5747
Format ascites
Immunogen Purified recombinant fragment of FAK expressed in E. Coli.
Form Liquid
Storage buffer ascites, 0.03% sodium azide
Temperatura przechowywania Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Shipping temperature Ice
Wprowadź szczegóły

Select the article for which you are looking for Certificate

Podaj numer partii. To pole jest obowiązkowe. Wpisz "X" dla VWR Chemicals, jeśli nr partii nie jest znany.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

albo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Opis Numer katalogowy Availability Jednostka Twoja Cena Price Per Ilość