Drukuj

Anti-SRA1 Mouse Monoclonal Antibody [clone: 7H1G1]

Dostawca: Thermo Fisher Scientific
PIEAMA515288EA 0 PLN
PIEAMA515288
Anti-SRA1 Mouse Monoclonal Antibody [clone: 7H1G1]
Przeciwciała
MA5-15288 targets SRA in IHC and WB applications and shows reactivity with Human samples. The MA5-15288 immunogen is purified recombinant fragment of SRA expressed in E. Coli. This gene is involved in transcriptional coactivation by steroid receptors. There is currently data suggesting this gene encodes both a non-coding RNA that functions as part of a ribonucleoprotein complex and a protein coding mRNA. Increased expression of both the transcript and the protein is associated with cancer.

Type: Primary
Antigen: SRA1
Clonality: Monoclonal
Clone: 7H1G1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR