MA5-15295 targets Lck in WB applications and shows reactivity with Human samples. The MA5-15295 immunogen is purified recombinant fragment of human Lck expressed in E. Coli. MA5-15295 detects Lck which has a predicted molecular weight of approximately 56kDa. This gene is a member of the Src family of protein tyrosine kinases . The encoded protein is a key signaling molecule in the selection and maturation of developing T-cells. It contains N-terminal sites for myristylation and palmitylation, a PTK domain, and SH2 and SH3 domains which are involved in mediating protein-protein interactions with phosphotyrosine-containing and proline-rich motifs, respectively. The protein localizes to the plasma membrane and pericentrosomal vesicles, and binds to cell surface receptors, including CD4 and CD8, and other signaling molecules. Multiple alternatively spliced variants, encoding the same protein, have been described.
Type: Primary
Antigen: LCK
Clonality: Monoclonal
Clone: 8E5F9, 4B2C7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human