Keep my session open?
Ending In 
The session is expired
Twoja sesja wygasła. Zostałeś wylogowany dla własnego bezpieczeństwa.
Chcesz zalogować się jeszcze raz?

Anti-SNCG Mouse Monoclonal Antibody [clone: 1H10D2]

Dostawca: Thermo Scientific
MA5-15296 targets SNCG (breast cancer-specific protein 1) in IHC and WB applications and shows reactivity with Human samples. The MA5-15296 immunogen is purified recombinant fragment of SNCG expressed in E. Coli. This gene encodes a member of the synuclein family of proteins which are believed to be involved in the pathogenesis of neurodegenerative diseases. Mutations in this gene have also been associated with breast tumor development.

Type: Primary
Antigen: SNCG
Clonality: Monoclonal
Clone: 1H10D2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human




 

ZAMÓW

Rozwiń / Zwiń
Rozmiar Ilość Numer katalogowy VWR Ilość
100 ug 100 µG PIEAMA515296 Blocked
Blocked Ten produkt jest zablokowany. Prosimy, skontaktuj się z Działem Obsługi Klienta, aby uzyskać więcej informacji.
Wygeneruj kody kreskowe Format kodu kreskowego: 
Wygeneruj stronę katalogową PDF

SPECYFIKACJE

Antigen symbol SNCG
Antigen name Synuclein gamma
Reactivity Human
Conjugation Unconjugated
Clonality Monoclonal
Antibody type Primary
Clone 1H10D2
Host Mouse
Isotype IgG1
Western blot Yes
ImmunoChemistry Yes
Antigen synonyms SR, SNCG, BCSG1
Gene ID 6623
Immunogen Purified recombinant fragment of SNCG expressed in E. Coli.
Purification purified
Form Liquid
Storage buffer PBS, 0.03% sodium azide
Temperatura przechowywania Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Shipping temperature Ice
Wprowadź szczegóły

Select the article for which you are looking for Certificate

Podaj numer partii. To pole jest obowiązkowe. Wpisz "X" dla VWR Chemicals, jeśli nr partii nie jest znany.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

albo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Opis Numer katalogowy Availability Jednostka Twoja Cena Price Per Ilość