Drukuj

Anti-FGG Mouse Monoclonal Antibody [clone: 13B304]

Dostawca: United States Biological
USBI030226EA 2960 PLN
USBI030226
Anti-FGG Mouse Monoclonal Antibody [clone: 13B304]
Przeciwciała
Type: Primary
Antigen: FGG (fibrinogen gamma chain)
Clonality: Monoclonal
Clone: 13B304
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR