Drukuj

Anti-FGR Mouse Monoclonal Antibody [clone: 13B306]

Dostawca: United States Biological
USBI030228EA 2850 PLN
USBI030228
Anti-FGR Mouse Monoclonal Antibody [clone: 13B306]
Przeciwciała
Type: Primary
Antigen: FGR (feline Gardner-Rasheed sarcoma viral oncogene homolog)
Clonality: Monoclonal
Clone: 13B306
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR