Drukuj

Anti-FBLN5 Mouse Monoclonal Antibody [clone: 13B307]

Dostawca: United States Biological
USBI030229EA 2850 PLN
USBI030229
Anti-FBLN5 Mouse Monoclonal Antibody [clone: 13B307]
Przeciwciała
Type: Primary
Antigen: FBLN5 (fibulin 5)
Clonality: Monoclonal
Clone: 13B307
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR