Drukuj

Anti-FBLN5 Mouse Monoclonal Antibody [clone: 13B308]

Dostawca: United States Biological
USBI030230EA 1970 PLN
USBI030230
Anti-FBLN5 Mouse Monoclonal Antibody [clone: 13B308]
Przeciwciała
Type: Primary
Antigen: FBLN5 (fibulin 5)
Clonality: Monoclonal
Clone: 13B308
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR