Drukuj
Anti-Flag Mouse Monoclonal Antibody [clone: 13B309]
Dostawca: United States Biological
USBI030231EA 2540 PLN
USBI030231
Anti-Flag Mouse Monoclonal Antibody [clone: 13B309]
Przeciwciała
Type: Tag
Antigen: Flag
Clonality: Monoclonal
Clone: 13B309
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Zobacz ceny