Drukuj

Anti-FOXA2 Mouse Monoclonal Antibody [clone: 13B316]

Dostawca: United States Biological
USBI030238EA 2640 PLN
USBI030238
Anti-FOXA2 Mouse Monoclonal Antibody [clone: 13B316]
Przeciwciała
Type: Primary
Antigen: FOXA2 (forkhead box A2)
Clonality: Monoclonal
Clone: 13B316
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR