Drukuj

Anti-FOXD3 Mouse Monoclonal Antibody [clone: 13B318]

Dostawca: United States Biological
USBI030240EA 2640 PLN
USBI030240
Anti-FOXD3 Mouse Monoclonal Antibody [clone: 13B318]
Przeciwciała
Type: Primary
Antigen: FOXD3 (forkhead box D3)
Clonality: Monoclonal
Clone: 13B318
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey, Mouse
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR