Drukuj
Anti-FOXP2 Mouse Monoclonal Antibody [clone: 13B322]
Dostawca: United States Biological
USBI030244EA 2960 PLN
USBI030244
Anti-FOXP2 Mouse Monoclonal Antibody [clone: 13B322]
Przeciwciała
Type: Primary
Antigen: FOXP2 (forkhead box P2)
Clonality: Monoclonal
Clone: 13B322
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human