Drukuj

Anti-FYN Mouse Monoclonal Antibody [clone: 13B328]

Dostawca: United States Biological
USBI030250EA 2960 PLN
USBI030250
Anti-FYN Mouse Monoclonal Antibody [clone: 13B328]
Przeciwciała
Type: Primary
Antigen: FYN (FYN oncogene related to SRC, FGR, YES)
Clonality: Monoclonal
Clone: 13B328
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR