Drukuj

Anti-FZD5 Mouse Monoclonal Antibody [clone: 13B330]

Dostawca: United States Biological
USBI030252EA 2960 PLN
USBI030252
Anti-FZD5 Mouse Monoclonal Antibody [clone: 13B330]
Przeciwciała
Type: Primary
Antigen: FZD5 (frizzled class receptor 5)
Clonality: Monoclonal
Clone: 13B330
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR