Drukuj

Anti-G6PD Mouse Monoclonal Antibody [clone: 13B332]

Dostawca: United States Biological
USBI030254EA 2960 PLN
USBI030254
Anti-G6PD Mouse Monoclonal Antibody [clone: 13B332]
Przeciwciała
Type: Primary
Antigen: G6PD (glucose-6-phosphate dehydrogenase)
Clonality: Monoclonal
Clone: 13B332
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR