Drukuj

Anti-GATA1 Mouse Monoclonal Antibody [clone: 13B336]

Dostawca: United States Biological
USBI030258EA 2710 PLN
USBI030258
Anti-GATA1 Mouse Monoclonal Antibody [clone: 13B336]
Przeciwciała
Type: Primary
Antigen: GATA1 (GATA binding protein 1 (globin transcription factor 1))
Clonality: Monoclonal
Clone: 13B336
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR