Drukuj

Anti-GATA5 Mouse Monoclonal Antibody [clone: 13B341]

Dostawca: United States Biological
USBI030263EA 2960 PLN
USBI030263
Anti-GATA5 Mouse Monoclonal Antibody [clone: 13B341]
Przeciwciała
Type: Primary
Antigen: GATA5 (GATA binding protein 5)
Clonality: Monoclonal
Clone: 13B341
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR