Drukuj

Anti-GSTM1 Mouse Monoclonal Antibody [clone: 13B360]

Dostawca: United States Biological
USBI030282EA 2960 PLN
USBI030282
Anti-GSTM1 Mouse Monoclonal Antibody [clone: 13B360]
Przeciwciała
Type: Primary
Antigen: GSTM1 (glutathione S-transferase mu 1)
Clonality: Monoclonal
Clone: 13B360
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR