Drukuj

Anti-GTF2B Mouse Monoclonal Antibody [clone: 13B363]

Dostawca: United States Biological
USBI030285EA 2710 PLN
USBI030285
Anti-GTF2B Mouse Monoclonal Antibody [clone: 13B363]
Przeciwciała
Type: Primary
Antigen: GTF2B (general transcription factor IIB)
Clonality: Monoclonal
Clone: 13B363
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR