Drukuj

Anti-HAS3 Mouse Monoclonal Antibody [clone: 13B367]

Dostawca: United States Biological
USBI030289EA 2960 PLN
USBI030289
Anti-HAS3 Mouse Monoclonal Antibody [clone: 13B367]
Przeciwciała
Type: Primary
Antigen: HAS3 (hyaluronan synthase 3)
Clonality: Monoclonal
Clone: 13B367
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR